Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005723

LC/MS Analysis of Polypeptide Drugs on Ascentis® Express RP-Amide

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase: [A] 10 mM ammonium acetate in water, pH 4.2; [B] 10 mM ammonium acetate in 90:10 (v/v) acetonitrile:water, pH 4.7
gradient: held at 20% B for 0.1 min; 20 to 80% B in 2.9 min; 80 to 20% B in 0.01 min; held at 20% B for 0.99 min
flow rate: 0.3 mL/min
column temp.: 35 °C
detector: TOF, ESI(+), m/z 100-2500
injection: 3 μL
sample: 1 μL/mL in 10mM ammonium formate, 80:20, acetonitrile:water

suitability

application for HPLC

application(s)

life science and biopharma

Analysis Note

This application demonstrates the suitability of the Ascentis Express RP Amide for the analysis of polypeptide drugs.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Oxytocin
    [deamino-Cys1, D-Arg8]-Vasopressin acetate salt hydrate

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service