Skip to Content
MilliporeSigma
All Photos(1)

Documents

G005454

SFC Analysis of Aminoglutethimide Enantiomers on Astec® Cellulose DMP

application for SFC

Sign Into View Organizational & Contract Pricing


About This Item

test parameters

column: Astec Cellulose DMP, 25 cm x 4.6 mm I.D., 5 μm particles (51099AST)
mobile phase: [A] CO2; [B] methanol containing 0.1% DEA in CO2; (75:25, A:B)
flow rate: 3 mL/min
pressure: 1740 psi (120 bar)
detector: UV, 230 nm
injection: 5 μL
sample: 2 mg/mL in methanol

suitability

application for SFC

application(s)

pharmaceutical (small molecule)

Analysis Note

This application demonstrates the ability of the Astec® Cellulose DMP column to separate Aminoglutethimide enantiomers via Supercritical Fluid Chromatography. DMPC-derivatized cellulose is widely used for chiral separations by SFC, both analytical and prep. The Astec® Cellulose DMP works well in SFC mode, providing rapid separations with excellent selectivity.

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    DL-Aminoglutethimide

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service