G004733
HPLC Analysis of Ritalinic Acid and Ritalin (Methylphenidate) Enantiomers on Astec® CHIROBIOTIC® V2 with T2 Guard Column
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: CHIROBIOTIC T2 guard column, 2 cm x 4 mm I.D., 5 μm particles (16100AST (requires holder 21150AST))
column: CHIROBIOTIC V2, 15 cm x 4.6 mm I.D., 5 μm particles (15023AST)
mobile phase: [A] 20 mM ammonium acetate, pH 4.1; [B] methanol; (7:93, A:B)
flow rate: 1 mL/min
column temp.: 25 °C
detector: UV, 230 nm
injection: 2 μL
sample: 1 mg/mL in methanol
suitability
application for HPLC
application(s)
forensics and toxicology
pharmaceutical (small molecule)
Legal Information
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service