Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G004631

HPLC Analysis of Methylphenobarbital (Mephobarbital) Enantiomers on Astec® CHIROBIOTIC® V

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase: [A] ethanol; [B] hexane; (25:75, A:B)
flow rate: 1 mL/min
column temp.: 25 °C
detector: UV, 205 nm
injection: 10 μL
sample: 1 mg/mL in methanol

suitability

application for HPLC

application(s)

pharmaceutical (small molecule)

Analysis Note

Methylphenobarbital (also known as mephobarbital and mephobarbitone) is a barbiturate derivative used primarily as an anticonvulsant, but also as a sedative and anxiolytic. It is a chiral compound and the enantiomers have been shown to exhibit different bioactivity, one narcotic, one convulsant, neither of which is seen in the racemate. Shown here is the rapid, efficient separation of methylphenobarbital enantiomers on an Astec CHIROBIOTIC V column. Highest grade HPLC solvents were used to supply low background interference and low particulate contamination for robust, trouble-free operation. A Sigma-Aldrich USP reference standard provided reliable identification and quantification.

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Mephobarbital, United States Pharmacopeia (USP) Reference Standard

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service