Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FFA57305

Supelco

SPME fiber assembly polyacrylate (PA)

df 85 μm(PA, for use with multi fiber exchanger, needle size 24 ga

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.21

material

fused silica fiber
plain white hub

needle size

24 ga

packaging

pack of 3
pkg of 3 ×  ea

df

85 μm (PA)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PA coating

application(s)

food and beverages

compatibility

for analyte group polar semivolatiles (MW 80-300)
for use with multi fiber exchanger

Looking for similar products? Visit Product Comparison Guide

Other Notes

Although we no longer distribute SPME-FFA products andaccessories, these products are available and distributed exclusively byChromline. Please visit the Chromline websiteor contact [email protected] for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)
Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)

Related Content

SPME fast-fit fiber assemblies simplify method development for solid-phase microextraction with easy installation and color-coded labels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service