Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F2053

Supelco

3-Hydroxycarbofuran

analytical standard

Synonym(s):

Carbofuran-3-hydroxy

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO4
CAS Number:
Molecular Weight:
237.25
Beilstein:
1383873
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 50 mg

manufacturer/tradename

Chem Service, Inc.

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

SMILES string

CNC(=O)Oc1cccc2C(O)C(C)(C)Oc12

InChI

1S/C12H15NO4/c1-12(2)10(14)7-5-4-6-8(9(7)17-12)16-11(15)13-3/h4-6,10,14H,1-3H3,(H,13,15)

InChI key

RHSUJRQZTQNSLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service