Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES5464

ChromegaChiral Chiral CCC, 10 Micron HPLC Column

10 μm particle size, L × I.D. 10 cm × 3.0 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

material

stainless steel column

Quality Level

product line

ChromegaChiral

manufacturer/tradename

ES Industries 123351-CCC

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

10 cm × 3.0 mm

matrix

fully porous particle

matrix active group

3 chloro-4 methylphenylcarbamate and 3,5-dichlorophenylcarbamate phase

particle size

10 μm

pore size

1000 Å pore size

operating pH

2-8

separation technique

chiral
supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

ChromegaChiral CCC is a modified cellulose including the combination of 3-chloro-4 methylphenylcarbamate and 3,5-dichloro- phenylcarbamate bonding groups coated on high purity, high performance spherical silica particles. This combination of bonded groups stabilizes the solubility of coated phase making for a durable phase similar to other widely used coated phases. The use of cellulose modified with chlorinated phenyl groups provides for the separation for many previously unresolved/poorly resolved chiral mixtures by providing unique separation characteristics.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChromegaChiral is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service