Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES5342

GreenSep Pyridyl Amide SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 5 cm × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52

material

stainless steel column

product line

GreenSep

manufacturer/tradename

ES Industries 112191-GS-PYA

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

5 cm × 2.1 mm

matrix

fully porous particle

matrix active group

pyridyl amide phase

particle size

3 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

The type of chemicals separated on a GreenSep Pyridyl Amide column would normally require the addition of TFA or an amine to the mobile phase as a peak shape modifier; however GreenSep Pyridyl Amide does not require this addition. Ideal for chemicals that contain both basic amine and acidic groups, as shown in the chromatogram below, GreenSep Pyridyl Amide provides flexibility for the SFC chromatographer with mobile phase composition and fraction collection greatly simplified without the use of amino additives.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service