Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES5152

GreenSep Cyano SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 25 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52

material

stainless steel column

product line

GreenSep

manufacturer/tradename

ES Industries 155191-GS-CN

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

matrix

fully porous particle

matrix active group

cyano phase

particle size

3 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase
supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

GreenSep Cyano is a high surface area Cyano bonded material designed for SFC resulting in a high surface area loading.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service