Skip to Content
MilliporeSigma
All Photos(1)

Documents

ERMAE003

Supelco

Tetramethylurea

ERM®, certified reference material

Synonym(s):

1,1,3,3-Tetramethylurea, TMU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCON(CH3)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
773898
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

form

liquid

manufacturer/tradename

JRC

refractive index

n20/D 1.451 (lit.)

bp

177 °C (lit.)
62-63 °C/12 mmHg (lit.)

mp

−1 °C (lit.)

density

0.968 g/mL at 20 °C (lit.)

SMILES string

CN(C)C(=O)N(C)C

InChI

1S/C5H12N2O/c1-6(2)5(8)7(3)4/h1-4H3

InChI key

AVQQQNCBBIEMEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please click on the below product number:
ERMAE003

Legal Information

ERM is a registered trademark of European Commission

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service