Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMB2154

Supelco

Protein (Nitrogen)

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

manufacturer/tradename

Elemental Microanalysis Limited

application(s)

food and beverages

format

matrix material

General description

Protein (Nitrogen), an organic analytical standard (OAS), consists of a homogenous batch of protein (casein) standard for use as a routine working micro-analytical standard for the determination of nitrogen. This OAS should be stored between 20°C to 25°C and should be kept tightly sealed away from light and moisture. It is non-hygroscopic under normal conditions and can be used without preliminary drying.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Elemental Analysis: Nitrogen

Matrix Group: Nuts, Soy, Edible Oils and Fats

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service