Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM96709

Supelco

Oleamide

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Ole, cis-9,10-Octadecenoamide, cis-9-Octadecenamide, Oleic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H35NO
CAS Number:
Molecular Weight:
281.48
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(N)=O

InChI

1S/C18H35NO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H2,19,20)/b10-9-

InChI key

FATBGEAMYMYZAF-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.
This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Biochem/physiol Actions

Sleep-inducing brain lipid, which allosterically modulates GABAA receptors and potentiates 5-HT7 serotonin receptor responses. Selective endogenous agonist of rat and human CB1 cannabinoid receptor.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service