Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM80832

Supelco

Cyprodinil Metabolite CGA 304075

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4-(4-Cyclopropyl-6-methylpyrimidin-2-ylamino)phenol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$180.00

$180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$180.00

About This Item

Empirical Formula (Hill Notation):
C14H15N3O
CAS Number:
Molecular Weight:
241.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$180.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

N(c2nc(cc(n2)C)C3CC3)c1ccc(cc1)O

InChI

1S/C14H15N3O/c1-9-8-13(10-2-3-10)17-14(15-9)16-11-4-6-12(18)7-5-11/h4-8,10,18H,2-3H2,1H3,(H,15,16,17)

InChI key

LBDZGKRFRHHISM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Cyprodinil Metabolite CGA 304075 is a para-phenol derivative of the anilinopyrimidine fungicide cyprodinil. Although it is formed as a minor degradation product of cyprodinil in plants, it occurs as its major metabolite in animals. Therefore, the residue definition of cyprodinil for its risk assessment and monitoring in foods of animal origin is taken as its sum with CGA 304075 (free and conjugated form). Cyprodinil is approved for its use in the European Union (EU) but due to the lack of a feeding study for the analysis of metabolite CGA 304075, MRLs for cyprodinil in animal commodities have not been proposed yet.
Cyprodinil has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conclusion regarding the peer review of the pesticide risk assessment of the active substance cyprodinil
European Food Safety Authority (EFSA)
EFSA Journal, 4(1), 51r-51r (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service