Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM36186

Supelco

Paraoxon-ethyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

O,O-Diethyl O-(4-nitrophenyl) phosphate, Diethyl p-nitrophenyl phosphate, Paraoxon, Paraoxon-ethyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OP(O)(OC2H5)2
CAS Number:
Molecular Weight:
275.20
Beilstein:
1915526
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.51 (lit.)

density

1.274 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

−20°C

SMILES string

CCOP(=O)(OCC)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C10H14NO6P/c1-3-15-18(14,16-4-2)17-10-7-5-9(6-8-10)11(12)13/h5-8H,3-4H2,1-2H3

InChI key

WYMSBXTXOHUIGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

Paraoxon-ethyl is a metabolite of parathion which is used as a non-systemic insecticide and acaricide to control sucking and chewing insects in cereals, fruit, citrus, vines, hops, vegetables, ornamentals, cotton and field crops. Paraoxon-ethyl is one of the most potent acetylcholinesterase-inhibing insecticides available. Under Reg. (EC) No 1107/2009 parathion is not approved for it′s use in the European Union as a plant protection product. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Biochem/physiol Actions

Potent irreversible acetylcholinesterase inhibitor

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service