Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM33887

Supelco

Desmethyl-formamido-pirimicarb

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

5,6-Dimethyl-2-(N-formylmethylamino)-4-pyrimidinyl dimethylcarbamate

Sign Into View Organizational & Contract Pricing

Select a Size

CRM33887-10MG
$264.00

$264.00


Please contact Customer Service for Availability


Select a Size

Change View
CRM33887-10MG
$264.00

About This Item

Empirical Formula (Hill Notation):
C11H16N4O3
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
41116107

$264.00


Please contact Customer Service for Availability

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

environmental

format

neat

storage temp.

−20°C

SMILES string

CN(C)C(=O)Oc1nc(nc(C)c1C)N(C)C=O

InChI

1S/C11H16N4O3/c1-7-8(2)12-10(15(5)6-16)13-9(7)18-11(17)14(3)4/h6H,1-5H3

InChI key

GDEAMEURJBBCOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Desmethyl-formamido-pirimicarb is a metabolite of pirimicarb. Pirimicarb is an carbamate insecticide for aphid control in a wide range of crops, such as wheat, potatoes, fruits, cotton tobacco. As per regulation (EC) No 1107/2009 pirimicarb is approved for its use as plant protection product in the European Union (EU). Maximum residue levels (MRLs) are set according toe Reg. (EU) 2016/71 for various products of plant and animal origin for pirimicarb from 0.01 to 15 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service