Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM32098

Supelco

Meptyldinocap

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(RS)-2-(1-Methylheptyl)-4,6-dinitrophenyl crotonate, 2-(1-Methylheptyl)-4,6-dinitrophenyl (2E)-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N2O6
CAS Number:
Molecular Weight:
364.39
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CCCCCCC(C)c1cc(cc(c1OC(=O)\C=C\C)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C18H24N2O6/c1-4-6-7-8-10-13(3)15-11-14(19(22)23)12-16(20(24)25)18(15)26-17(21)9-5-2/h5,9,11-13H,4,6-8,10H2,1-3H3/b9-5+

InChI key

NIOPZPCMRQGZCE-WEVVVXLNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Environmental
Meptyldinocap is a protectant, curative and eradicant dinitrophenol fungicide, a single isomer of dinocap and active against powdery mildew mainly on fruit and other crops. It has a low aqueous solubility, it is volatile and, based on its chemical properties, is unlikely to leach to groundwater. As per the regulation (EC) No 1107/2009 it is approved for use as plant protection product in the European Union. Maximum residuel levels (MRLs) have been set for meptyldinocap (sum of 2,4 DNOPC and 2,4 DNOP expressed as meptyldinocap) according Reg. (EU) No 2021/1864 for various products of plant and animal origin from 0.01 - 0.5 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service