Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

cis/trans FAME Column Performance Mix

analytical standard, 2.5 mg/mL in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

analytical standard

packaging

pkg of 1 × 1 mL (40495-U)
pkg of 10 × 1 mL (4M0495-U)

concentration

2.5 mg/mL in methylene chloride

storage temp.

2-8°C

General description

Qualitative mix. Contains many of the C18:1 postional cis/trans isomers of great importantance to the food industry.

Data sheet includes representative chromatogram and instructions on how to use the mix to monitor for GC system and column performance changes.

Application

Useful for monitoring unexpected changes in the performance of cyanosilicone capillary columns as suggested in method AOCS Ce 1h-05, Determination of cis-, trans-, Saturated, Monounsaturated and Polyunsaturated Fatty Acids in Vegetable or Non-ruminant Animal Oils and Fats by Capillary GLC, 2005.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service