Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96047

Supelco

1,4-Cyclohexanedicarboxylic acid

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$84.30

$84.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$84.30

About This Item

Linear Formula:
C6H10(CO2H)2
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21

$84.30


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% ((mixture of isomers), GC)

technique(s)

HPLC: suitable

mp

164-167 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)C1CCC(CC1)C(O)=O

InChI

1S/C8H12O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)

InChI key

PXGZQGDTEZPERC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service