Skip to Content
MilliporeSigma
All Photos(1)

Documents

95644

Supelco

Cyflufenamid metabolite 149-F1

PESTANAL®, analytical standard

Synonym(s):

2,3-Difluoro-6-(trifluoromethyl)benzenecarboximidamide, 2,3-Difluoro-6-(trifluoromethyl)benzamidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5F5N2
CAS Number:
Molecular Weight:
224.13
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥95.0% (HPLC)

format

neat

storage temp.

−20°C

General description

Cyflufenamid is a preventative and curative amidoxine fungicide for the control of powdery mildew in cereals and other crops. It has a low aqueous solubility and is non-volatile. It is approved according Reg. (EC) No 1107/2009 for its use as a plant protection product in the European Union (EU).

Application

Environmental

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service