Skip to Content
MilliporeSigma
All Photos(1)

Documents

93222

Supelco

Benzovindiflupyr

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-[9-(Dichloromethylene)-1,2,3,4-tetrahydro-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15Cl2F2N3O
CAS Number:
Molecular Weight:
398.23
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

InChI

1S/C18H15Cl2F2N3O/c1-25-7-11(15(24-25)17(21)22)18(26)23-12-4-2-3-8-9-5-6-10(13(8)12)14(9)16(19)20/h2-4,7,9-10,17H,5-6H2,1H3,(H,23,26)/t9-,10+/m1/s1

InChI key

CCCGEKHKTPTUHJ-ZJUUUORDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application


  • Advanced disease control in agriculture: Benzovindiflupyr fungicide demonstrates significant efficacy in controlling sclerotinia diseases in crops, contributing to sustainable agricultural practices and enhancing crop yield and quality. Its mode of action involves inhibiting the succinate dehydrogenase pathway, crucial for fungal respiration (Cui et al., 2024).

  • In planta resistance mechanisms: In-depth studies into the resistance mechanisms of fungi to Benzovindiflupyr help in understanding how genetic mutations affect fungicide efficacy. This research is pivotal in managing resistance and developing next-generation fungicides (Matsuzaki et al., 2024).

  • Role in integrated pest management: Benzovindiflupyr is utilized in integrated pest management programs to control a wide range of fungal pathogens in diverse agricultural settings. Its broad-spectrum activity makes it a valuable tool for managing disease in crops such as cereals, fruits, and vegetables, contributing to global food security (Wong and Quesada-Ocampo, 2024).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service