Skip to Content
MilliporeSigma
All Photos(1)

Documents

92266

Supelco

p-Phenylenediamine

analytical standard

Synonym(s):

1,4-Benzenediamine, 1,4-Diaminobenzene, 1,4-Phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2
CAS Number:
Molecular Weight:
108.14
Beilstein:
742029
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

grade

analytical standard

Quality Level

vapor density

3.7 (vs air)

vapor pressure

1.08 mmHg ( 100 °C)

description

anti-fade reagent

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

267 °C (lit.)

mp

138-143 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(N)cc1

InChI

1S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4H,7-8H2

InChI key

CBCKQZAAMUWICA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 1 Oral

Target Organs

Kidney,Heart,Musculo-skeletal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service