Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90648

Supelco

Oleamide-13C18

analytical standard

Synonym(s):

Oleic acid amide-13C18, cis-9,10-Octadecenoamide-13C18, cis-9-Octadecenamide-13C18

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$433.00

$433.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$433.00

About This Item

Empirical Formula (Hill Notation):
13C18H35NO
Molecular Weight:
299.34
UNSPSC Code:
41116107
NACRES:
NA.24

$433.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service