Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

844465

TOYOPEARL® Hexyl-650C Bulk Media

bottle of 25 mL, phase C6 (hexyl), 100 μm particle size

Synonym(s):

TOYOPEARL® Hexyl-650 HIC Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$331.00

$331.00


Please contact Customer Service for Availability


Select a Size

Change View
25 ML
$331.00

About This Item

UNSPSC Code:
41106500

$331.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 44465

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

C6 (hexyl) phase

particle size

100 μm

operating pH

1-13

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl Hexyl-650 resin has the highest hydrophobicity of the HIC ligands offered by TBL. It is used for very hydrophilic proteins or where a low salt elution environment is required. It is useful as an early capture step or for interresinte purification.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service