Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

843400

TOYOPEARL® AFFIPAK ACT Sampler (AF-Epoxy, Tresyl-650M)

phase AF-Epoxy, Tresyl-650M

, bottle of 2 × 5 g, 65 μm particle size

Synonym(s):

TOYOPEARL® AFFIPAK Activated Media Sampler

Sign Into View Organizational & Contract Pricing

Select a Size

2 X 5 G
$501.00

$501.00


Please contact Customer Service for Availability


Select a Size

Change View
2 X 5 G
$501.00

About This Item

UNSPSC Code:
41115700

$501.00


Please contact Customer Service for Availability

description

Phase filter (Various)

product line

TOYOPEARL®

form

dry, free flowing powder

feature

Molecular weight range (Excl. limit: 5,000,000 Da)

packaging

bottle of 2 × 5 g

manufacturer/tradename

Tosoh 43400

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

particle size

65 μm

pore size

1000 Å

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service