Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

843203

TOYOPEARL® CM-650M Bulk Media

phase carboxymethyl, bottle of 100 mL, 65 μm particle size

Synonym(s):

TOYOPEARL® CM-650 Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$392.00

$392.00


Please contact Customer Service for Availability


Select a Size

Change View
100 ML
$392.00

About This Item

UNSPSC Code:
41115700

$392.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 100 mL

manufacturer/tradename

Tosoh 43203

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

carboxymethyl phase

particle size

65 μm

operating pH

2-13

capacity

40 g/L±10 adsorption capacity (lysozyme)

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl CM-650 resin is a weak cation exchanger for biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service