Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

822866

TOYOPEARL GigaCap® DEAE-650M Bulk Media

bottle of 250 mL, phase DEAE (diethylaminoethyl), 75 μm particle size

Synonym(s):

TOYOPEARL GigaCap® DEAE-650M Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$1,290.00

$1,290.00


Please contact Customer Service for Availability


Select a Size

Change View
250 ML
$1,290.00

About This Item

UNSPSC Code:
41115700

$1,290.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

feature

counter ion Cl-

packaging

bottle of 250 mL

manufacturer/tradename

Tosoh 22866

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

DEAE (diethylaminoethyl) phase

particle size

75 μm

operating pH

2-13

capacity

>156 g/L adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl GigaCap DEAE-650M resin is a very high capacity anion exchange resin. Toyopearl HW-65 is the polymeric base bead chemically modified to provide a greater number of anionic binding sites resulting in increased binding capacity. Toyopearl GigaCap DEAE-650M exhibits typical dynamic binding capacities (DBC) approaching 170 g/L for bovine serum albumin (BSA) and has the ability to separate proteins up to 96 g/L.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation
Toyopearl GigaCap is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service