Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821454

TSKgel® ODS-100Z (5 µm) HPLC Columns

L x I.D. 1.5 cm × 3.2 mm, C18 (octadecyl) phase

Sign Into View Organizational & Contract Pricing

Select a Size

3 PKG
$535.00

$535.00


Please contact Customer Service for Availability


Select a Size

Change View
3 PKG
$535.00

About This Item

UNSPSC Code:
41115711

$535.00


Please contact Customer Service for Availability

Product Name

TSKgel® ODS-100Z Guard Cartridge, phase C18 (octadecyl), L × I.D. 1.5 cm × 3.2 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh 21454

extent of labeling

20% Carbon, encapped with trimethylsilane

parameter

0-100% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1.5 cm × 3.2 mm

surface area

450 m2/g

matrix

spherical silica

matrix active group

C18 (octadecyl) phase

particle size

5 μm

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The bonded phase of TSKgel ODS-100Z columns is prepared by reacting the silica surface with a difunctional octadecylsilane reagent, followed by repeated endcapping with a monofunctional trimethylsilane reagent. The resulting TSKgel ODS-100Z columns contain a high density monomeric C18 bonded phase for maximum retention and selectivity of small molecular weight compounds and minimal secondary interaction with residual silanol groups.
This general purpose reversed phase column is the workhorse for analysis of small molecular weight compounds in life science applications. Lot-to-lot reproducibility stands out.

Physical form

Shipped in acetonitrile.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service