Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82020-U

Hamilton® PRP-X100 (10 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, PEEK hardware

Synonym(s):

Hamilton PRP-X100 reverse phase column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,420.00

$1,420.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$1,420.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$1,420.00


Please contact Customer Service for Availability

Product Name

Hamilton PRP-X100 HPLC Column, 10 μm particle size, L × I.D. 25 cm × 4.6 mm, PEEK hardware

material

PEEK column

manufacturer/tradename

Hamilton

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

ammonium ion, quaternary phase

particle size

10 μm

pore size

100 Å

operating pH

1-14

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

The Hamilton PRP-X100, polymeric HPLC columns used for the separation of anions in ion chromatography.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service