Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

818546

TSKgel® SuperQ-5PW (20) Bulk Media

bottle of 1 L, phase ammonium ion, quaternary, 20 μm particle size

Synonym(s):

TSKgel® SuperQ-5PW Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$12,760.00

$12,760.00


Please contact Customer Service for Availability


Select a Size

Change View
1 L
$12,760.00

About This Item

UNSPSC Code:
41115700

$12,760.00


Please contact Customer Service for Availability

Agency

suitable for USP L23

product line

TSKgel®

form

slurry

feature

counter ion Cl-

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 18546

parameter

20 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

ammonium ion, quaternary phase

particle size

20 μm

operating pH

2-13

capacity

70 g/L±18 adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel SP-5PW media is a strong cation exchanger for biomolecule purification. Its small particle size makes it an excellent choice for polishing and high performance resolution steps. It has the same selectivity as the larger particle Toyopearl SP-650 resin.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

103.1 °F

Flash Point(C)

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service