Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

818344

TSKgel® Alpha-M (13 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Alpha Columns

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,820.00

$2,820.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$2,820.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

$2,820.00


Please contact Customer Service for Availability

Product Name

TSKgel® Alpha-M HPLC Column, phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 13 μm particle size

material

stainless steel hardware

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 18344

parameter

≤0.5 M salt concentration
0-100% polar organic solvent
1 mL/min flow rate
20 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

13 μm

pore size

mixed

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Alpha columns are composed of spherical, hydrophilic methacrylate beads. Unlike TSKgel PW-type columns, which are stable at most to a 50% organic mixed with water, TSKgel Alpha-type columns are stable in a wide variety of organic solvents at concentrations up to 100%. The main application area for TSKgel Alpha columns is the analysis of polymers that are soluble in polar organic solvents. Examples include cellulose derivatives, polyimide, and sodium dodecylsulfate (all in DMF with 10mmol/L DMF), cleansing gel in methanol, and degree of saponification of polyvinylalcohol in HFIP. The mixed-bed column, TSKgel Alpha-M, has an extended linear calibration range, and is suitable for samples with a broad MW distribution as well as samples with unknown molecular weight.
TSKgel Alpha-M columns are so-called mixed-bed columns with an extended linear calibration range. They are packed with rigid, porous, polymer beads which show minimal shrinkage or swelling in different solvent systems. They are best used for the separation of polymers that are soluble in polar organic solvents such as methanol, ethanol, DMF, THF, HFIP, DMSO, 2-propanol, and acetonitrile.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service