Skip to Content
MilliporeSigma
All Photos(1)

Documents

818206

TSKgel® HPLC Columns Hardware and Accessories

for P/N 818207, pkg of 1 ea

Synonym(s):

Holder for TSKgel® Super Series Guardfilters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

TSKgel® Hardware and Accessories, for P/N 818207, pkg of 1 ea

material

stainless steel

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 18206

technique(s)

HPLC: suitable

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Protect your 4.6 mm ID TSKgel Super Series column (Super-Octyl, Super-ODS and Super-Phenyl) from plugging by installing a guard filter in front of the analytical column. The Holder for Super Series Guardfilters requires 0.4 cm L x 4 mm ID Guardfilters (PN 818207) that need to be purchased separately.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service