Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

817227

TOYOPEARL® SuperQ-650M Bulk Media

phase ammonium ion, quaternary, bottle of 250 mL, 65 μm particle size

Synonym(s):

TOYOPEARL® SuperQ-650 Anion Exchange Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501

description

SuperQ-650M

product line

TOYOPEARL®

form

liquid

feature

counter ion Cl-

packaging

bottle of 250 mL

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

methacrylate

matrix active group

ammonium ion, quaternary phase

particle size

65 μm

operating pH

2-12

capacity

0.20-0.30 meq/mL ion exchange capacity
130 g/L±25 adsorption capacity (BSA)
143 mg/mL binding capacity (bovine serum albumin)

compatibility

mode of use strong anion exchange chromatography

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl SuperQ-650 resin is a strong anion exchanger for biomolecule purification. It can be used for high throughput capture, interresinte purification, and polishing process steps. If more powerful resolution is needed the smaller particle TSKgel SuperQ-5PW resin should be used. Selectivity remains the same.

Specifications

Clean in place with 0.1 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service