Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

816654

TSKgel® OApak-P (10 µm) HPLC Columns

L x I.D. 4 cm × 6 mm, sulphonic acid phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® OApak-P Guard Column, phase sulphonic acid, L × I.D. 4 cm × 6 mm, 10 μm particle size

material

stainless steel

product line

TSKgel®

feature

counter ion H+

manufacturer/tradename

Tosoh 16654

parameter

≤20% organic solvent
0.8 mL/min flow rate
10-60 °C temperature
70 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

4 cm × 6 mm

matrix

hydroxylated methacrylic polymer

matrix active group

sulphonic acid phase

particle size

10 μm

operating pH

2-12

separation technique

cation exchange

General description

TSKgel® OApak-P guard columns is used in conjunction with the TSKgel OApak-A analytical column. The TSKgel OApak-P guard column contains a strong cation exchanger for the removal of dissociated strong acids under the isocratic eluent conditions of 0.75mM H2SO4.Applications include: organic acids in fruit juices, wine, beer, coffee, and salt solutions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Science Slam panel: Leading gene therapy developers discuss commercialization challenges and the importance of robust process development plans.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service