Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814714

TSKgel® SP-5PW (20) Bulk Media

bottle of 250 mL, phase sulfonic acid, 20 μm particle size

Synonym(s):

TSKgel® SP-5PW Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$4,160.00

$4,160.00


Please contact Customer Service for Availability


Select a Size

Change View
250 ML
$4,160.00

About This Item

UNSPSC Code:
41115700

$4,160.00


Please contact Customer Service for Availability

product line

TSKgel®

form

slurry

feature

counter ion Na+

packaging

bottle of 250 mL

manufacturer/tradename

Tosoh 14714

parameter

20 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

sulfonic acid phase

particle size

20 μm

operating pH

2-13

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel SP-5PW resin is a strong cation exchanger for biomolecule purification. Its small particle size makes it an excellent choice for polishing and high performance resolution steps. It has the same selectivity as the larger particle Toyopearl SP-650 resin.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

103.1 °F

Flash Point(C)

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service