Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

814026

Supelco

TOYOPEARL® QAE-550C Bulk Media

phase ammonium ion, quaternary, bottle of 250 mL, 100 μm particle size

Synonym(s):

TOYOPEARL® QAE-550C Anion Exchange Media

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$599.00

$599.00


Please contact Customer Service for Availability


Select a Size

Change View
250 ML
$599.00

About This Item

UNSPSC Code:
47101511

$599.00


Please contact Customer Service for Availability

description

QAE-550C

product line

TOYOPEARL®

form

slurry

feature

counter ion Cl-

packaging

bottle of 250 mL

parameter

3 bar max. pressure

technique(s)

HPLC: suitable

matrix

methacrylate

matrix active group

ammonium ion, quaternary phase

particle size

100 μm

operating pH

2-10

capacity

0.37 meq/mL ion exchange capacity
70 g/L±10 adsorption capacity (BSA)
70 mg/mL binding capacity (bovine serum albumin)

compatibility

mode of use strong anion exchange chromatography

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl QAE-550 resin is a strong anion exchanger useful for purification of proteins up to 100kD. It is used for high throughput capture of feedstocks.

Application

TOYOPEARL® QAE-550C is used in separation media and ion exchange resins.

Specifications

Clean in place with 0.1 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Staby et al.
Journal of chromatography. A, 908(1-2), 149-161 (2001-02-24)
A comparative study was performed on strong anion exchangers to investigate the pH dependence, titration curves, efficiency, binding strength, particle size distribution, and static and dynamic capacity of the chromatographic resins. The resins tested included Q Sepharose XL, UNO Q-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service