Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808804

TSKgel® Phenyl-5PW (10 µm) HPLC Columns

L x I.D. 7.5 cm × 8 mm, phenyl phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,590.00

$2,590.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$2,590.00

About This Item

UNSPSC Code:
41115711

$2,590.00


Please contact Customer Service for Availability

Product Name

TSKgel® Phenyl-5PW HPLC Column, phase phenyl, L × I.D. 7.5 cm × 8 mm, 10 μm particle size

material

glass

product line

TSKgel®

manufacturer/tradename

Tosoh 8804

parameter

≤4 M salt concentration
≤50% organic solvent
1.2 mL/min flow rate
10-50 °C temperature
20 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 8 mm

matrix

hydroxylated methacrylic polymer

matrix active group

phenyl phase

particle size

10 μm

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

The most hydrophobic among the three TSKgel® HIC columns, TSKgel Phenyl-5PW is an excellent choice to screen for the selectivity, retention and recovery of most biomolecules. Featuring a 1000Ã… pore size in a polymethacrylate base material, the Phenyl-5PW column is stable in either acid or caustic cleaning regimens and provides excellent access to larger molecules with low diffusion coefficients. Stainless steel is the standard column format for TSKgel Phenyl-5PW. Glass and PEEK column housings are available for samples that may adsorb to stainless steel. The alternative column formats are also suitable for general applications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service