Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808002

TOYOPEARL® AF-Amino-650M Bulk Media

phase amine-activated, bottle of 25 mL, 65 μm particle size

Synonym(s):

TOYOPEARL® AF-Amino-650M Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$379.00

$379.00


Please contact Customer Service for Availability


Select a Size

Change View
25 ML
$379.00

About This Item

UNSPSC Code:
41115700

$379.00


Please contact Customer Service for Availability

description

AF-Amino-650M

product line

TOYOPEARL®

form

liquid

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh TOYOPEARL® AF, 08002

extent of labeling

100 μmol/mL±30 ligand density

parameter

3 bar max. pressure

technique(s)

HPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

amine-activated phase

particle size

65 μm

operating pH

2-13

separation technique

anion exchange

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Amino-650 resin is a reactive resin used for the coupling of specific ligands for affinity chromatography. Ligands are immobilized by either peptide bond formation or reductive amination through their respective carboxylate or aldehyde groups.

Application

TOYOPEARL® Affinity Media are used in affinity chromatography, protein chromatography and activated/functionalized matrices.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Knör et al.
Journal of thrombosis and haemostasis : JTH, 6(3), 470-477 (2008-01-16)
Hemophilia A is currently treated by infusions of the coagulation factor (F) VIII, of which production and purification remain a challenging task. Current purification procedures using immunoaffinity chromatography are cumbersome, expensive, and suffer from the instability of the applied antibody
S Yamauchi et al.
The Journal of biological chemistry, 274(4), 2456-2463 (1999-01-16)
Chondroitin 4-sulfotransferase, which transfers sulfate from 3'-phosphoadenosine 5'-phosphosulfate to position 4 of N-acetylgalactosamine in chondroitin, was purified 1900-fold to apparent homogeneity with 6.1% yield from the serum-free culture medium of rat chondrosarcoma cells by affinity chromatography on heparin-Sepharose CL-6B, Matrex
Zhenzhen Wen et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(20), 1732-1740 (2011-05-17)
In order to improve its stability, immobilized Concanavalin A (Con A) on Toyopearl adsorbents was conjugated with monomethoxy poly(ethylene glycol) succinimidyl propionate (mPEG-SPA) with different molecular weight. A colorimetric method using ninhydrin is proposed to determine the degree of PEGylation;

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service