Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807166

TSKgel® QAE-2SW (5 µm) HPLC Columns

L x I.D. 25 cm × 4.6 mm, ammonium ion, quaternary phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,540.00

$2,540.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$2,540.00

About This Item

UNSPSC Code:
41115711

$2,540.00


Please contact Customer Service for Availability

Product Name

TSKgel® QAE-2SW HPLC Column, phase ammonium ion, quaternary, L × I.D. 25 cm × 4.6 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L14

product line

TSKgel®

feature

counter ion H2PO4-

manufacturer/tradename

Tosoh 07166

parameter

≤1 M salt concentration
0-100% organic solvent
1.0 mL/min flow rate
10-45 °C temperature
150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

matrix

spherical silica

matrix active group

ammonium ion, quaternary phase

particle size

5 μm

operating pH

2.0-7.5

separation technique

anion exchange

General description

TSKgel® QAE-2SW columns are packed with 5μm porous spherical silica beads which are surface modified with a strong anion exchanger. They are used for the separation and analysis of peptides, nucleotides, nucleosides and other biologically active molecules.

Physical form

Shipped in methanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service