Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

807165

TSKgel® SP-2SW (5 µm) HPLC Column

L x I.D. 25 cm × 4.6 mm, sulfonic acid phase

Synonym(s):

TSKgel® Cation Exchange HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,550.00

$2,550.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$2,550.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

$2,550.00


Please contact Customer Service for Availability

Product Name

TSKgel® SP-2SW HPLC Column, phase sulfonic acid, L × I.D. 25 cm × 4.6 mm, 5 μm particle size

material

stainless steel hardware

Agency

suitable for USP L52
suitable for USP L9

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh SP-2SW, 07165

parameter

≤1 M salt concentration
0-100% organic solvent
1.0 mL/min flow rate
10-45 °C temperature
150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

matrix

silica particle platform

matrix active group

sulfonic acid phase

particle size

5 μm

pore size

125 Å

operating pH

2.0-7.5

separation technique

cation exchange

General description

TSKgel® SP-2SW columns are packed with 5 micron, high efficiency, spherical silica beads which are surface modified with a strong cation exchanger. They are used for the separation and analysis of small proteins, peptides, nucleosides, and other biologically active molecules.

Application

TSKgel® SP-2SW HPLC column may be used as a cation exchanger column in the determination of histamine in biological materials by high-performance liquid chromatography (HPLC).[1]

Physical form

Shipped in methanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of plasma and brain histamine without previous purification of biological samples: cation-exchange chromatography coupled with post-column derivatization fluorometry.
Watanabe T.
Journal of Chromatography. B, Biomedical Sciences and Applications, 344 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service