Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

806758

TSKgel® PW-Type (17 µm) HPLC Columns

L x I.D. 7.5 cm × 21.5 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® PW-Type Guard Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,350.00

$2,350.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$2,350.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

$2,350.00


Please contact Customer Service for Availability

Product Name

TSKgel® PW-Type Guard Column, phase hydroxylated methacrylate, L × I.D. 7.5 cm × 21.5 mm, 17 μm particle size

material

stainless steel

Agency

suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 06758

parameter

≤0.5 M salt concentration
≤20% organic solvent
8 mL/min flow rate
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 21.5 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

17 μm

pore size

200 Å

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Guard Column for 21.5mmID TSKgel G2500-G3000PW columns, 17μm

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service