Skip to Content
MilliporeSigma
All Photos(1)

Documents

77655

Supelco

4-Chlororesorcinol

analytical standard

Synonym(s):

1,3-Dihydroxy-4-chlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OH)2
CAS Number:
Molecular Weight:
144.56
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

147 °C/18 mmHg (lit.)

mp

106-108 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Oc1ccc(Cl)c(O)c1

InChI

1S/C6H5ClO2/c7-5-2-1-4(8)3-6(5)9/h1-3,8-9H

InChI key

JQVAPEJNIZULEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service