Skip to Content
MilliporeSigma
All Photos(1)

Documents

72646

Supelco

4-Allyloxy-2-hydroxybenzophenone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2OC6H3(OH)COC6H5
CAS Number:
Molecular Weight:
254.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

67-70 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Oc1cc(OCC=C)ccc1C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-2-10-19-13-8-9-14(15(17)11-13)16(18)12-6-4-3-5-7-12/h2-9,11,17H,1,10H2

InChI key

GVZIBGFELWPEOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service