Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

713-1192

GC Analysis of a 7-Component Organochlorine Pesticide Mix in Drinking Water on a 5% Phenyl Column after SPE using Supelclean ENVI-18

suitable for GC, application for SPE

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

test parameters

sample preparation: SPE (Solid Phase Extraction)
sample/matrix: drinking water spiked with pesticides
SPE tube/cartridge: Supelclean ENVI-18, 500 mg/6 mL (57064)
condition: 2 x 6 mL hexane:ethyl ether (1:1); 6 mL methanol; 6 mL DI water
sample addition: 250 mL water sample at 10 mL/min
drying: 10 min with vacuum
elution: 2 x 15 mL hexane:ethyl ether (1:1)
eluate post-treatment: concentrate to 2 mL with nitrogen stream
column: 5% Phenyl Polysiloxane, 30 m × 0.25 mm I.D., 0.25 μm (Supelco equivalent, Equity-5, 28089-U)
oven: 150 °C (2 min), 10 °C/min to 275 °C
detector: ECD, 310 °C
carrier gas: helium
injection: 1 μL

suitability

application for SPE

application(s)

environmental
food and beverages

Legal Information

ENVI is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    Hexachlorobenzene, analytical standard
    Aldrin, PESTANAL®, analytical standard
    Endrin, PESTANAL®, analytical standard
    Heptachlor, analytical standard
    Heptachlor epoxide, PESTANAL®, analytical standard, isomer B
    γ-BHC, certified reference material, 1000 μg/mL in methanol
    Methoxychlor, analytical standard
    Lindane, PESTANAL®, analytical standard
    See All (8)

SPE tube or plate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service