Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69027

Supelco

Bis(2-ethylhexyl) sebacate

analytical standard

Synonym(s):

Di(2-ethylhexyl) sebacate, Sebacic acid di(2-ethylhexyl) ester, ‘Dioctyl’ sebacate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$80.60

$80.60


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
$80.60

About This Item

Linear Formula:
[-(CH2)4CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
426.67
Beilstein:
1806504
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.24

$80.60


Please contact Customer Service for Availability

grade

analytical standard

Quality Level

Assay

≥97%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.450 (lit.)

bp

212 °C/1 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

InChI

1S/C26H50O4/c1-5-9-17-23(7-3)21-29-25(27)19-15-13-11-12-14-16-20-26(28)30-22-24(8-4)18-10-6-2/h23-24H,5-22H2,1-4H3

InChI key

VJHINFRRDQUWOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

awg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service