Skip to Content
MilliporeSigma
All Photos(1)

Documents

64823

Supelco

Flat Bottom Flask

size 300 mL

Synonym(s):

FLAT BOTTOM FLASK,300ML,24/40, EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

flat bottom glass flask

Quality Level

size

large
300 mL

Joint

joint ST/NS flask

application(s)

food and beverages

compatibility

for use with liquid/liquid extractors

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service