Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63468

Supelco

Silica gel on TLC Al foils

silica gel 60 matrix, L × W 10 cm × 20 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
L × W:
10 cm × 20 cm
layer thickness:
0.20 mm
surface area:
~500 m2/g
pore size:
0.75 mL/g pore volume
60 Å mean pore size

material

aluminum support
silica gel 60 matrix

technique(s)

thin layer chromatography (TLC): suitable

L × W

10 cm × 20 cm

layer thickness

0.20 mm

surface area

~500 m2/g

particle size

5-17 μm

pore size

0.75 mL/g pore volume
60 Å mean pore size

Looking for similar products? Visit Product Comparison Guide

General description

  • Outstanding wettability (even for 100% aqueous visualisation reagents)
  • Easy and precise cutting (no flaking of silica)
  • Excellent separation efficiency and reproducibility from batch to batch
polymer binder, is stable in almost all organic solvents and resistant towards aggressive visualisation reagents and aqueous eluents

Other Notes

this product will be discontinued, please find the alternative product here :
105553

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service