Key Documents
58468
Supelco
Dynaseal Fittings
pkg of 10 ea, Kel-F ferrule
Synonym(s):
Kel-F FERRULES,1/16" PK/10
Select a Size
10 EA
$66.70
$66.70
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
10 EA
$66.70
About This Item
UNSPSC Code:
41115700
$66.70
Please contact Customer Service for Availability
Request a Bulk Order
material
Kel-F ferrule
packaging
pkg of 10 ea
fitting
1/16 in. Dynaseal ferrule for 1/16 in. O.D. tubing
Features and Benefits
Dynaseal hand-tight fittings will seal against pressures to over 7000psi/490kg/cm². Each fitting consists of a reusable nut, collet, and ferrule. The polymer ferrule will not deform stainless steel seats and can be reused. Except for the fitting for Rheodyne valves, these fittings are compatible with all 1/16 in. female compression-type fittings now used on HPLC columns.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)