Skip to Content
MilliporeSigma
All Photos(1)

Documents

58424

Supelco

LC-NH2 Test Mix

in acetonitrile: water (25:75), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

in acetonitrile: water (25:75)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

compatibility

use to QA LC-NH2 columns

format

multi-component solution

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Analyte

Description

    D-(−)-Fructose 25 mg/mL
    α-D-Glucose 25 mg/mL
    Lactose 25 mg/mL
    Maltose 25 mg/mL
    Sucrose 25 mg/mL

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service