Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

581392-U

Supelco

Supelguard HPLC Guard Cartridge Holder

Stand-Alone, for use with Supelguard cartridges (1 cm L. x 21.2 mm I.D.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Supelguard Guard Cartridge Holder, Stand-Alone, for use with Supelguard cartridges (1 cm L. x 21.2 mm I.D.)

technique(s)

HPLC: suitable

compatibility

for use with Supelguard cartridges (1 cm L. x 21.2 mm I.D.)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service