Skip to Content
MilliporeSigma
All Photos(1)

Documents

58060-U

Nylon 66 Filter Membranes

pore size 0.20 μm, diam. 47 mm, pkg of 50

Synonym(s):

MEMBRANES NYLON 66 0.2UM PK/50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

Nylon 66

packaging

pkg of 50

diam.

47 mm

pore size

0.20 μm

Looking for similar products? Visit Product Comparison Guide

Application

Nylon 66 filter membranes is used for sample extraction in the analysis of polycyclic musks (HHCB, AHTN, ADBI, AHDI, ATII) and a metabolite of HHCB (HHCB-lactone) in sewage sludge samples using high-resolution gas-chromatography with ion-impact-selective ion monitoring mass spectrometry (HRGC-EI-SIM-MS).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concentrations and specific loads of polycyclic musks in sewage sludge originating from a monitoring network in Switzerland.
Kupper T, et al.
Chemosphere, 54(8), 1111-1120 (2004)
J O Thostenson et al.
Water research, 140, 191-199 (2018-05-02)
Electrochemical disinfection (ECD) has become an important blackwater disinfection technology. ECD is a promising solution for the 2 billion people without access to conventional sanitation practices and in areas deficient in basic utilities (e.g., sewers, electricity, waste treatment). Here, we
Mengge Chen et al.
Artificial cells, nanomedicine, and biotechnology, 48(1), 824-833 (2020-05-28)
Zinc pyrithione (ZPT) is widely used as an antimicrobial. Zinc is a necessary trace element of the human whose homeostasis associated with several cancers. However, the anticancer effect of increased Zinc in ovarian cancer is still unclear. This study focussed
Parshuram J Sonawane et al.
PloS one, 6(1), e16661-e16661 (2011-02-10)
3-Hydroxy-3-methylglutaryl-coenzyme A [HMG-CoA] reductase gene (Hmgcr) is a susceptibility gene for essential hypertension. Sequencing of the Hmgcr locus in genetically hypertensive BPH (blood pressure high), genetically hypotensive BPL (blood pressure low) and genetically normotensive BPN (blood pressure normal) mice yielded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service